Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Trypsin Inhibitor, Corn

Catalog No. 6503451MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-034-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-034-51MG Supplier MilliporeSigma™ Supplier No. 6503451MG
Add to Cart
Add to Cart

The Trypsin Inhibitor, Corn, also referenced under CAS 9035-81-8, controls the biological activity of Trypsin. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Specific, reversible inhibitor of trypsin. Also inhibits human Factor XIIa. May function in host resistance against fungal pathogens in crops.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 30 mM NaCl, 20 mM Tris-HCl, pH 8.2.
Description Single band by SDS-PAGE
Target Proteins trypsin
Formula Weight 14000g/mol
Solubility Aqueous buffers
Content And Storage Avoid Freeze-Thaw Cycles
Form Liquid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.