Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ U-73343

Catalog No. 6620415MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-204-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-204-15MG Supplier MilliporeSigma™ Supplier No. 6620415MG
Add to Cart
Add to Cart

A cell-permeable analog of U-73122 that acts as a very weak inhibitor of phospholipase C. Suitable as a negative control.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by TLC
Target Proteins Negative control of U-73122 in the inhibition of phospholipase C
CAS 142878-12-4
Molecular Formula C29H42N2O3
Formula Weight 466.7g/mol
Solubility CHCl3, DMSO, or EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.