Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ U0124

Catalog No. 6620061MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-200-61MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-200-61MG Supplier MilliporeSigma™ Supplier No. 6620061MG
Add to Cart
Add to Cart

A useful negative control for MEK inhibitors U0125 and U0126.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by HPLC
Target Proteins MEK1
CAS 1140528-25-1
Molecular Formula C8H10N4S2
Formula Weight 226.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.