Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Ubiquitin Isopeptidase Inhibitor I, G5

Catalog No. 66212510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-212-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-212-510MG Supplier MilliporeSigma™ Supplier No. 66212510MG
Add to Cart
Add to Cart

The Ubiquitin Isopeptidase Inhibitor I, G5, also referenced under CAS 108477-18-5, controls the biological activity of Ubiquitin Isopeptidase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 97% by Elemental analysis
Target Proteins Ubiquitin isopeptidase activity
CAS 108477-18-5
Molecular Formula C19H14N2O7S
Formula Weight 414.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.