Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ UCN-01

Catalog No. 539644500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-964-4500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-964-4500UG Supplier MilliporeSigma™ Supplier No. 539644500UG
Add to Cart
Add to Cart

A cell-permeable Staurosporine derived anticancer agent that reversibly and ATP-competitively inhibits several protein kinases.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Tan
Description 95% by HPLC
Target Proteins PKCα, PKCβ, PKCγ, PKCδ PKCε, Chk1, Cdc25C-associated protein kinase 1, Cdk1, PAK4, Cdk5/p25 and Chk2, PDK1, lck, MAPKAP kinase-2, Akt, GSK-3β and PKA
CAS 112953-11-4
Molecular Formula C28H26N4O4
Formula Weight 482.5g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.