Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Urea Transporter B Inhibitor, UTBinh-14

Catalog No. 66612510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-612-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-612-510MG Supplier MilliporeSigma™ Supplier No. 66612510MG
Add to Cart
Add to Cart

A cell-permeable triazolothienopyrimidine compound that acts as a reversible urea transporter B-selective inhibitor (IC 50 =

A cell-permeable triazolothienopyrimidine compound that acts as a reversible urea transporter B-selective inhibitor (IC 50 = 10.3 and 25.1nM against urea influx using human and mouse erythrocytes, respectively) by targeting a UT-B intracellular site in a urea-competitive manner, exhibiting little effect toward UT-A1, UT-A3, AQP1, CFTR, or TMEM16A channel activities. Shown to effectively decrease maximum urinary concentration and increase urination volume from antidiuretic dDAVP-treated wild-type, but not UT-B -/- , mice (300μg/300μl/mouse i.p.) in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins urea transporter B
Molecular Formula C20H17N5O2S3
Formula Weight 455.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.