Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Urokinase Substrate III, Fluorogenic

Catalog No. 67215925MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-215-925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-215-925MG Supplier MilliporeSigma™ Supplier No. 67215925MG
Add to Cart
Add to Cart

Suitable for the quantitative direct fluorescent assay of urokinase.

Peptide Sequence: Z-Gly-Gly-Arg-AMC, HCl
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Description 95% by HPLC
Target Proteins Quantitative direct fluorescent assay of urokinase
Molecular Formula C28H33N7O7·HCl
Formula Weight 616.1g/mol
Solubility EtOH or H2O
Content And Storage Protect from moisture
Form Lyophilized
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.