Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ V-ATPase Inhibitor, KM91104

Catalog No. 67299910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-299-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-299-910MG Supplier MilliporeSigma™ Supplier No. 67299910MG
Add to Cart
Add to Cart

A cell-permeable, non-macrolide small molecule V-ATPase inhibitor that specifically targets the a3-b2 subunit protein protein interation of V-ATPase (IC50 o

A cell-permeable, non-macrolide small molecule V-ATPase inhibitor that specifically targets the a3-b2 subunit protein protein interation of V-ATPase (IC50 of 2.3μM), in vitro . It is also shown to inhibit osteoclast resorption (IC50 of 1.2μM) on both synthetic hydroxyapatite surfaces and dentin slices in an in vitro osteoclast model, RANKL-differentiated RAW 264.7 cells, without significantly affecting RAW 264.7 cell viability or RANKL-mediated osteoclast differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description >95% by HPLC
Molecular Formula C14H12N2O4
Formula Weight 272.08g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.