Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Packaged Under Inert Gas
- Peptide Sequence: cyclic(D-Phe-Pro-Gln-Ile-Met-Arg-Ile-Lys-Pro-His-Gln-Gly-Gln-His-Ile-Gly-Glu)
Specifications
Specifications
| Color | White |
| Formulation | Supplied as a trifluoroacetate salt. |
| Description | 97% by HPLC |
| Target Proteins | Blocks the binding of VEGF165 to its receptors |
| Molecular Formula | C90H140N28O22S |
| Formula Weight | 1998.3g/mol |
| Solubility | DMSO or H2O |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Lyophilized |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?