Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ VEGF Inhibitor, V1

Catalog No. 6764951MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-649-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-649-51MG Supplier MilliporeSigma™ Supplier No. 6764951MG
Add to Cart
Add to Cart

The VEGF Inhibitor, V1 controls the biological activity of VEGF. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: H-Ala-Thr-Trp-Leu-Pro-Pro-Arg-OH
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Specifically competes against VEGF165 binding to recombinant NRP-1
Molecular Formula C40H61N11O9
Formula Weight 840g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.