Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Veratridine

Catalog No. 6769505MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-695-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-695-05MG Supplier MilliporeSigma™ Supplier No. 6769505MG
Add to Cart
Add to Cart

A Na+ channel opener.

A Na+ channel opener. Increases intracellular Ca2+ levels without affecting Na+ - Ca2+ exchange. Acts at neurotoxin receptor site 2 and preferentially binds to activated Na+ channels causing persistent activation. Induces cell death in bovine chromaffin cells.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Description 98% by TLC
Target Proteins Na+ channel
CAS 71-62-5
Molecular Formula C36H51NO11
Formula Weight 673.8g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.