Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Vigabatrin

Catalog No. 5059890001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-598-90001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-598-90001 Supplier MilliporeSigma™ Supplier No. 5059890001
Add to Cart
Add to Cart

A selective, irreversible inhibitor of GABA transaminase with antiepileptic properties

A selective, irreversible inhibitor of GABA transaminase with antiepileptic properties. Covalently binds to the active site of GABA tansaminase to inactivate it in a dose-dependent manner and increases extracellular GABA levels in the brain. Rapidly absorbed following oral administration and is shown to inhibit [3H]GABA uptake across the apical membrane of Caco-2 cell monolayers in a concentration-dependent manner (IC50 = 2.9 mM)
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by HPLC
Target Proteins GABA Transaminase
Molecular Formula C6H11NO2
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.