Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Vinblastine Sulfate

Catalog No. 67717510MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-717-510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-717-510MG Supplier MilliporeSigma™ Supplier No. 67717510MG
Add to Cart
Add to Cart

Antitumor alkaloid drug.

Antitumor alkaloid drug. Induces apoptosis in cultured hepatocytes and human lymphoma cells. Affects interaction of tubulin with microtubule-associated proteins, specifically Tau and MAP2. Depolymerizes microtubules. Increases frequency of micronucleated erythrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Interaction of tubulin with microtubule-associated proteins, specifically Tau and MAP2
CAS 143-67-9
Molecular Formula C46H58N4O9·H2SO4
Formula Weight 909.1g/mol
Solubility H2O
Content And Storage Protect from Light, Protect from Moisture
Form Crystalline Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.