Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ WAY-100635 Maleate

Catalog No. 5081460001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-814-60001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-814-60001 Supplier MilliporeSigma™ Supplier No. 5081460001
Add to Cart
Add to Cart

A selective 5-HT1A receptor antagonist (pKi

A selective 5-HT1A receptor antagonist (pKi = 7.9-9.2). WAY-100635 has been reported to have good inhibitory action centrally and peripherally (1mg/kg dose in rats) but has low intrinsic effects, making it a silent 5-HT1A receptor antagonist. WAY-100635 displaced specific binding of the 5-HT1A radioligand, [3H]8-OH-DPAT (8-hydroxy-2-(di-n-propylamino)tetralin), to rat hippocampal membranes with a pIC50 of 8.87. This represented a greater than 100-fold selectivity relative to binding at other 5-HT receptor subtypes and major neurotransmitter receptor, reuptake and ion channel sites. WAY-100635 has been used in a variety of vertebrate species in vivo and in vitro .
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 97% by HPLC
Target Proteins 5-HT1A
Molecular Formula C25H34N4O2·C4H4O4
Solubility Warm H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.