Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Wee1/Chk1 Inhibitor

Catalog No. 6816371MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-163-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-163-71MG Supplier MilliporeSigma™ Supplier No. 6816371MG

The Wee1/Chk1 Inhibitor, also referenced under CAS 1177150-89-8, controls the biological activity of Wee1/Chk1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 95% by HPLC
Target Proteins Wee1 and Chk1
CAS 1177150-89-8
Molecular Formula C20H12N2O3·H2O
Formula Weight 346.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.