Learn More
MilliporeSigma™ Calbiochem™ Western-Re-Probe™ Reagent
Catalog No. WB59100ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
WB59100ML 100 mL
1 options
Catalog No. WB59100ML Supplier MilliporeSigma™ Supplier No. WB59100ML
Add to Cart
Add to Cart
Enhances blotting capacity of proteins.
  • Validated for use in Immunoblotting
  • Dissociates remove antibodies from membrane-bound proteins without destroying antigenic binding capacity
  • For stripping chemiluminescent or radioisotopic signals from blots
  • Packaging: Glass Bottle
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL