Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ WHI-P180

Catalog No. 6815001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-150-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-150-01MG Supplier MilliporeSigma™ Supplier No. 6815001MG
Add to Cart
Add to Cart

A potent inhibitor of lgE-mediated mast cell responses to allergens in vitro and in vivo.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins Cdk2
Molecular Formula C16H15N3O3
Formula Weight 297.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.