Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ WRN Helicase Inhibitor, NSC 19630

Catalog No. 6816475MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-164-75MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-164-75MG Supplier MilliporeSigma™ Supplier No. 6816475MG
Add to Cart
Add to Cart

The WRN Helicase Inhibitor, NSC 19630, also referenced under CAS 72835-26-8, controls the biological activity of WRN Helicase. This small molecule/inhibitor is primarily used for Cell Signaling applications.

A cell-permeable maleimide compound selectively inhibits Werner syndrome WRN helicase activity (IC50 = 20μM) over its ATPase and exonuclease activities (19% and <10% inhibition, respectively, at 50μM), while exhibiting no effect against the helicase activity of two other human RecQ family DNA helicases (RECQ1 and Werner syndrome BLM), Fanconi anemia group J helicase FANCJ, as well as three other E. coli helicases, RecQ, UvrD, and DnaB. The WRN-dependent antiproliferation activity of MIRA-1/NSC 19630 in HeLa 1.2.11 cultures (0% vs. 95% inhibition with or without siRNA-mediated WRN-depletion, respectively, after 48 h 3μM treatment) is shown to be a direct result of apoptosis induction due to replication forks blockage (20-fold increase in PCNA foci staining after 72 h 2μM treatment), enhanced DSB (17-fold increase in γ-H2AX foci staining after 72 h 2μM treatment), and increased S-phase population (from 24% to 42% after 72 h 2μM treatment). In some cells, MIRA-1/NSC 19630 appears to also exert its apoptotic effect, at least in part, by restoring DNA-binding and transactivation function of some, but not all, p53 mutants.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by NMR
CAS 72835-26-8
Molecular Formula C8H9NO4
Formula Weight 183.2g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.