Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Xanthine

Catalog No. 68205GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-205-GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-205-GM Supplier MilliporeSigma™ Supplier No. 68205GM

Nitrogenous by-product of nucleoprotein metabolism

An intermediate in the degradation of adenosine monophosphate to uric acid
TRUSTED_SUSTAINABILITY

Specifications

CAS 69-89-6
Color White to slightly yellowish
Content And Storage Store at +15°C to +30°C
Formula Weight 152.1g/mol
Molecular Formula C5H4N4O2
Purity ≥99% by HPLC
Quantity 5 g
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.