Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Xestospongin C, Xestospongia sp.

Catalog No. 68216010UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-216-010UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-216-010UG Supplier MilliporeSigma™ Supplier No. 68216010UG
Add to Cart
Add to Cart

An oxaquinolizidine alkaloid isolated from a marine sponge.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 90% by HPLC
Target Proteins IP3-mediated Ca2+ release
Molecular Formula C28H50N2O2
Formula Weight 446.7g/mol
Solubility DMSO, EtOH, or MeOH
Content And Storage Protect from light
Form Solid
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.