Learn More
Norco Safety Calibration Gas: Air, Disposable, Hydrogen Sulphide 25 ppm, +/-5% Accuracy, Calibration Gas, 58 L Cylinder Capacity
Catalog No. NC3316210
Encompass
Portable gas detector calibration gases are precise, known mixtures of specified gases contained in a pressurized cylinder. They work with compatible testing equipment to confirm or adjust a detector's alert parameters.
Catalog No. NC3316210 Supplier Norco Safety Supplier No. NLBZ105325PA
Portable gas detector calibration gases are precise, known mixtures of specified gases contained in a pressurized cylinder. They work with compatible testing equipment to confirm or adjust a detector's alert parameters.