Learn More
Aat Bioquest Calbryte 590 potassium salt (5x50 ug)
SDP
Supplier:  Aat Bioquest 20706
Encompass_Preferred
The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G proteinquotcoupled receptors GPCRs and calcium channel targets. This catalog is for 5x50 ug.
Catalog No. 50-224-9890