Learn More
Sino Biological CALCA / CGRP Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 13933RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CALCA / CGRP (rh CALCA; Catalog#13933-H09E; P01258-1; Ala26-Asn141). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-8318