Learn More
Enzo Life Sciences Calcitonin gene related peptide monoclonal antibody (CD8) (100µl)
SDP
Supplier:  Enzo Life Sciences BMLCA11350100
Encompass_Preferred
Clone: CD8. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of C-terminal rat calcitonin gene-related peptide (CGRP). Species reactivity: Human Guinea pig. Applications: IHC. Purity: Partially purified.
Catalog No. 50-201-0354