Learn More
Enzo Life Sciences Calcitonin gene related peptide polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLCA11370100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of rat α-calcitonin gene-related peptide (CGRP). Source: Purified from sheep serum. Species reactivity: Human, Mouse, Rat, Rabbit. Applications: IHC. Purity: Ammonium sulphate precipitation. Formulation: Liquid. In PBS containing 0.1% sodium azide.
Catalog No. 50-201-0356