Learn More
Enzo Life Sciences Calcitonin gene related peptide polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLCA11340025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to a portion of rat α-calcitonin gene-related peptide (CGRP). Source: Whole rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: IHC. Formulation: Liquid. Antiserum containing 0.1% sodium azide.
Catalog No. 50-201-0353