Learn More
Medchemexpress LLC HY-D0040 100mg Medchemexpress, Calcein CAS:1461-15-0 Purity:>98%
SDP
Catalog No. 501933137
Encompass_Preferred
Medchemexpress, HY-D0040 100mg Calcein CAS:1461-15-0 Calcein is a fluorescent dye and self-quenching probe, used as an indicator of lipid vesicle leakage, and also as a complexometric indicator for titration of calcium ions with EDTA, and for fluorometric determination of calcium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3137 Supplier Medchemexpress LLC Supplier No. HYD0040100MG
Medchemexpress, HY-D0040 100mg Calcein CAS:1461-15-0 Calcein is a fluorescent dye and self-quenching probe, used as an indicator of lipid vesicle leakage, and also as a complexometric indicator for titration of calcium ions with EDTA, and for fluorometric determination of calcium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.