Learn More
AdipoGen Calcein-AM (1 mg)
SDP
Catalog No. 501490280
Encompass_Preferred
Calcein-AM (1 mg), AdipoGen Life Sciences. Chemical. CAS: 148504-34-1. Formula: C46H46N2O23. MW: 994.86. Synthetic. Calcein-AM is a non-fluorescent cell permeable and hydrophobic probe that, upon entering live cells, is cleaved by intracellular esterases, releasing the membrane-impermeable, hydrophilic, and intensely bright green fluorescent calcein. Better retained by viable cells than fluorescein, carboxyfluorescein, or BCECF, due to its low cytotoxicity. It can be used as an indicator of cell viability (dead cells lack cytoplasmic esterases), cell-cell communication, cytotoxicity or changes in intracellular calcium, fluoride, iron or mercury. Used as a cytosolic fluorophore in mitochondrial permeability studies to image pore transition. Viability assays using calcein are reliable and correlate well with the standard 51Cr-release assay. Spectral Data: λex 496nm; λem 516nm in 0.1M Tris pH 8.0.
Catalog No. 50-149-0280 Supplier AdipoGen Supplier No. CDXC0009M001
Calcein-AM (1 mg), AdipoGen Life Sciences. Chemical. CAS: 148504-34-1. Formula: C46H46N2O23. MW: 994.86. Synthetic. Calcein-AM is a non-fluorescent cell permeable and hydrophobic probe that, upon entering live cells, is cleaved by intracellular esterases, releasing the membrane-impermeable, hydrophilic, and intensely bright green fluorescent calcein. Better retained by viable cells than fluorescein, carboxyfluorescein, or BCECF, due to its low cytotoxicity. It can be used as an indicator of cell viability (dead cells lack cytoplasmic esterases), cell-cell communication, cytotoxicity or changes in intracellular calcium, fluoride, iron or mercury. Used as a cytosolic fluorophore in mitochondrial permeability studies to image pore transition. Viability assays using calcein are reliable and correlate well with the standard 51Cr-release assay. Spectral Data: λex 496nm; λem 516nm in 0.1M Tris pH 8.0.