Learn More
AnaSpec Calcein, AM 1mg, 148504-34-1
Catalog No. 50850334
Encompass
Calcein, AM is a cell-permeant and non-fluorescent compound that is widely used for determining cell viability. In live cells the non-fluorescent calcein, AM is hydrolyzed by intracellular esterases into the strongly green fluorescent anion calcein. The fluorescent calcein is well-retained in the cytoplasm in live cells. Hela cells stained with AnaSpec’s Calcein AM fluorescent dye. MW 994.9; (1 mg)
Catalog No. 50-850-334 Supplier AnaSpec Supplier No. AS89201
Calcein, AM is a cell-permeant and non-fluorescent compound that is widely used for determining cell viability. In live cells the non-fluorescent calcein, AM is hydrolyzed by intracellular esterases into the strongly green fluorescent anion calcein. The fluorescent calcein is well-retained in the cytoplasm in live cells. Hela cells stained with AnaSpec’s Calcein AM fluorescent dye. MW 994.9; (1 mg)