Learn More
Biotium Calcein AM, 20x50 ug
Supplier:  Biotium 800113
Encompass_Preferred
Calcein AM is a widely used green fluorescent cell marker. Calcein AM is membrane-permeant and thus can be introduced into cells via incubation. Once inside the cells, calcein AM is hydrolyzed by endogenous esterase into the highly negatively charged green fluorescent calcein, which is retained in the cytoplasm. Calcein AM is an excellent tool for the studies of cell membrane integrity and for cell tracing. Supplied in lyophilized form or as a solution in anhydrous DMSO at 4 mM (80011-1) or 1 mg/mL (1 mM) (80011-2). Calcein AM is also offered in our Cell Viability and Cytotoxicity Assay Kit. Properties: Ex/Em (calcein) = 494/517 nm (pH 8); Colorless solid soluble in DMSO; Store at -20°C and protect from light; C46H46N2O23; MW: 995
Catalog No. 50-196-4780