Learn More
Aat Bioquest Calcein AM CAS 148504-34-1 (1 mg)
SDP
Catalog No. 502250017
Encompass_Preferred
Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein. This catalog is for 1 mg.
This item is not returnable. View return policy
Catalog No. 50-225-0017 Supplier Aat Bioquest Supplier No. 22002
Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein. This catalog is for 1 mg.