Learn More
Aat Bioquest Calcein AM UltraPure grade CAS 148504-34-1 (1 mg)
SDP
Catalog No. 502250018
Encompass_Preferred
Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein. This catalog is for 1 mg.
This item is not returnable. View return policy
Catalog No. 50-225-0018 Supplier Aat Bioquest Supplier No. 22003
Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein. This catalog is for 1 mg.