Learn More
Specifications
Specifications
| For Use With (Application) | Cell Staining |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye |
| Content And Storage | Store at -20°C or lower and protect from light. |
| Label Type | Other Labels or Dyes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.