Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calcein-AM, Calbiochem™,

Catalog No. 2067001MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-670-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-670-01MG Supplier MilliporeSigma™ Supplier No. 2067001MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 148504-34-1
Molecular Formula C46H46N2O23
Molecular Weight (g/mol) 994.87
MDL Number MFCD05861516
InChI Key BQRGNLJZBFXNCZ-UHFFFAOYSA-N
Synonym Calcein-acetoxymethyl Ester, Diacetate, CAL-AM
IUPAC Name (acetyloxy)methyl 2-({2-[(acetyloxy)methoxy]-2-oxoethyl}({[3',6'-bis(acetyloxy)-7'-{[bis({2-[(acetyloxy)methoxy]-2-oxoethyl})amino]methyl}-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl]methyl})amino)acetate
SMILES CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)CC1=CC2=C(OC3=CC(OC(C)=O)=C(CN(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)C=C3C22OC(=O)C3=CC=CC=C23)C=C1OC(C)=O

Specifications

Packaging Glass Bottle
Color White
Quantity 1 mg
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.