Learn More
Apexbio Technology LLC Calcein AM Cell Viability Assay Kit 500T
SDP
Catalog No. 5000064305
Encompass_Preferred
The Calcein AM Cell Viability Assay Kit employs calcein AM a cell-permeable fluorescent probe for assessing viable cell populations Calcein AM readily crosses membranes due to its hydrophobic acetoxymethyl (AM) modification Within viable cells intracellular esterases cleave calcein AM releasing calcein a fluorescent molecule retained by healthy cells emitting green fluorescence proportional to viable cell numbers Cells lacking this enzymatic activity typically dead or compromised remain unstained This assay is commonly applied in studies evaluating cell proliferation cytotoxicity or cell survival under various experimental conditions Compared to assays such as MTT and CCK-8 the calcein AM method allows detection of smaller cell populations often down to approximately 50 cells and provides results within approximately 30 minutes after treatment
Catalog No. 50-000-64305 Supplier Apexbio Technology LLC Supplier No. K2231500
The Calcein AM Cell Viability Assay Kit employs calcein AM a cell-permeable fluorescent probe for assessing viable cell populations Calcein AM readily crosses membranes due to its hydrophobic acetoxymethyl (AM) modification Within viable cells intracellular esterases cleave calcein AM releasing calcein a fluorescent molecule retained by healthy cells emitting green fluorescence proportional to viable cell numbers Cells lacking this enzymatic activity typically dead or compromised remain unstained This assay is commonly applied in studies evaluating cell proliferation cytotoxicity or cell survival under various experimental conditions Compared to assays such as MTT and CCK-8 the calcein AM method allows detection of smaller cell populations often down to approximately 50 cells and provides results within approximately 30 minutes after treatment