Learn More
Cayman Chemical CalceIn Blue 25mg
SDP
Catalog No. 502984413
Encompass_Preferred
A membrane-impermeant fluorescent dye; ex/em maxima = 360/445 nm
Catalog No. 50-298-4413 Supplier Cayman Chemical Supplier No. 2063625 MG
A membrane-impermeant fluorescent dye; ex/em maxima = 360/445 nm