Learn More
Cayman Chemical CalceIn Blue AM 1mg
SDP
Catalog No. 502982332
Encompass_Preferred
A fluorogenic dye used to assess cell viability; ex/em maxima = 360/445 nm, respectively
Catalog No. 50-298-2332 Supplier Cayman Chemical Supplier No. 206371 MG
A fluorogenic dye used to assess cell viability; ex/em maxima = 360/445 nm, respectively