Learn More
Cayman Chemical CalceIn Deep Red1mg
SDP
Catalog No. 502982277
Encompass_Preferred
A membrane-impermeant fluorescent dye; ex/em maxima = 646/659 nm, respectively
Catalog No. 50-298-2277 Supplier Cayman Chemical Supplier No. 206341 MG
A membrane-impermeant fluorescent dye; ex/em maxima = 646/659 nm, respectively