Learn More
Cayman Chemical CalceIn Deep RedAM 1mg
SDP
Catalog No. 502991841
Encompass_Preferred
A fluorogenic probe; non-fluorescent but cell-permeant and is hydrolyzed to the fluorescent probe Calcein Deep Red™ that displays excitation/emission maxima of 643/663 nm, respectively
Catalog No. 50-299-1841 Supplier Cayman Chemical Supplier No. 305751 MG
A fluorogenic probe; non-fluorescent but cell-permeant and is hydrolyzed to the fluorescent probe Calcein Deep Red™ that displays excitation/emission maxima of 643/663 nm, respectively