Learn More
Cayman Chemical CalceIn Ornge sodIum salt 1mg
SDP
Catalog No. 502982966
Encompass_Preferred
A membrane-impermeant fluorescent dye; ex/em maxima = 525/550 nm, respectively
Catalog No. 50-298-2966 Supplier Cayman Chemical Supplier No. 206401 MG
A membrane-impermeant fluorescent dye; ex/em maxima = 525/550 nm, respectively