Learn More
Cayman Chemical CalceIn OrngeDIacetate 1mg
SDP
Catalog No. 502983569
Encompass_Preferred
A fluorogenic dye used to assess cell viability; ex/em = 525/550 nm, respectively
Catalog No. 50-298-3569 Supplier Cayman Chemical Supplier No. 206411 MG
A fluorogenic dye used to assess cell viability; ex/em = 525/550 nm, respectively