Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcein, Reagent grade, MP Biomedicals™

Catalog No. ICN19508701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19508701 1 g
ICN19508705 5 g
ICN19508710 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19508701 Supplier MP Biomedicals, Inc Supplier No. 0219508701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1461-15-0 | C30H26N2O13 | 622.539 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-15-0
Molecular Formula C30H26N2O13
Molecular Weight (g/mol) 622.539
MDL Number MFCD00005049
InChI Key DEGAKNSWVGKMLS-UHFFFAOYSA-N
Synonym Fluorescein-methyleneiminodiacetic acid, Fluorexon
PubChem CID 65079
ChEBI CHEBI:51903
IUPAC Name 2-[[7'-[[bis(carboxymethyl)amino]methyl]-3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-2'-yl]methyl-(carboxymethyl)amino]acetic acid
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)CN(CC(=O)O)CC(=O)O)O)O)CN(CC(=O)O)CC(=O)O

Specifications

Color Orange
Quantity 1 g
Physical Form Powder
Formula Weight 622.5
Grade Research
Chemical Name or Material Calcein
EINECSNumber 215-957-1
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.