Learn More
Cayman Chemical CalceIn Red sodIum salt 1mg
SDP
Catalog No. 502983983
Encompass_Preferred
A membrane-impermeant fluorescent dye; ex/em = 560/574 nm, respectively
Catalog No. 50-298-3983 Supplier Cayman Chemical Supplier No. 206331 MG
A membrane-impermeant fluorescent dye; ex/em = 560/574 nm, respectively