Learn More
Cayman Chemical Calcein Red™ AM, Purity: ≥90%, 1 mg
SDP
Catalog No. 502985570
Encompass_Preferred
A fluorogenic dye used to assess cell viability; ex/em = 560/574 nm, respectively
Catalog No. 50-298-5570 Supplier Cayman Chemical Supplier No. 206321 MG
A fluorogenic dye used to assess cell viability; ex/em = 560/574 nm, respectively