Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcein Sodium Salt (mixture of isomers) [for Fluorometric Determination of Ca], TCI America™
SDP

Catalog No. C00035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C00035G Supplier TCI America Supplier No. C00035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00076066
Synonym Fluorescein Bis(methyliminodiacetic Acid) Sodium Salt, Fluorexon Sodium Salt, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein Sodium Salt

Specifications

Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Calcein Sodium Salt (mixture of isomers) [for Fluorometric Determination of Ca]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.