Learn More
Cayman Chemical CalceIn tetraethyl ester 10mg
SDP
Catalog No. 502988861
Encompass_Preferred
An intermediate in the synthesis of calcein AM
Catalog No. 50-298-8861 Supplier Cayman Chemical Supplier No. 3075410 MG
An intermediate in the synthesis of calcein AM