Learn More
Medchemexpress LLC Calcein (tetraethyl ester) | 1170856-93-5 | >99.5% | 1 ML
SDP
Catalog No. 5000410972
Encompass_Preferred
Calcein tetraethyl ester is a fluorescent dye used in biology. It can be transported through the cellular membrane into live cells, which makes it useful for testing of cell viability and for short-term labeling of cells.
  • Fluorescent dye.
  • Can be transported through the cellular membrane.
  • Useful for testing cell viability.
  • Useful for short-term labeling of cells.
Catalog No. 50-004-10972 Supplier Medchemexpress LLC Supplier No. HYD083010MM/1ML
Calcein tetraethyl ester is a fluorescent dye used in biology. It can be transported through the cellular membrane into live cells, which makes it useful for testing of cell viability and for short-term labeling of cells.
  • Fluorescent dye.
  • Can be transported through the cellular membrane.
  • Useful for testing cell viability.
  • Useful for short-term labeling of cells.