Learn More
Medchemexpress LLC Calcein (tetraethyl ester) | 1170856-93-5 | 99.5% | 5 MG
SDP
Catalog No. 5000409636
Encompass_Preferred
Calcein tetraethyl ester, also known as Fluorexon tetraethyl ester, is a fluorescent dye utilized in biological applications. Its key characteristic is its ability to be transported through the cellular membrane into live cells. This property makes it valuable for assessing cell viability and for the short-term labeling of cells.
  • Effective for live cell studies: Its cell-permeable nature allows for direct application in living systems without damaging cell integrity.
  • Versatile in research: Can be used for various analyses including cell function, in vivo imaging, and cell staining.
Catalog No. 50-004-09636 Supplier Medchemexpress LLC Supplier No. HYD08305MG
Calcein tetraethyl ester, also known as Fluorexon tetraethyl ester, is a fluorescent dye utilized in biological applications. Its key characteristic is its ability to be transported through the cellular membrane into live cells. This property makes it valuable for assessing cell viability and for the short-term labeling of cells.
  • Effective for live cell studies: Its cell-permeable nature allows for direct application in living systems without damaging cell integrity.
  • Versatile in research: Can be used for various analyses including cell function, in vivo imaging, and cell staining.