Learn More
Cayman Chemical CalceIn tetraethyl ester 1mg
SDP
Catalog No. 502989705
Encompass_Preferred
An intermediate in the synthesis of calcein AM
Catalog No. 50-298-9705 Supplier Cayman Chemical Supplier No. 307541 MG
An intermediate in the synthesis of calcein AM