Learn More
Cayman Chemical CalceIn UltraBlue sodIM sa 1mg
SDP
Catalog No. 502984815
Encompass_Preferred
A membrane-impermeant fluorescent dye; ex/em maxima = 360/445 nm
Catalog No. 50-298-4815 Supplier Cayman Chemical Supplier No. 206381 MG
A membrane-impermeant fluorescent dye; ex/em maxima = 360/445 nm